Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KMT2B
Entrez Gene ID
718835
Gene Name
lysine methyltransferase 2B
Gene Aliases
MLL4
Chromosome Location
Chr.19: 35131201 - 35152046 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015123697.2 XP_014979183.2 - 7879 61
XM_028839234.1 XP_028695067.1 - 7615 61
XM_028839235.1 XP_028695068.1 - 7525 61
XM_028839236.1 XP_028695069.1 - 7759 61

View Full Product Details