Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP6V1G3
Entrez Gene ID
710372
Gene Name
ATPase H+ transporting V1 subunit G3
Gene Aliases
EGK_01818
Chromosome Location
Chr.1: 55939309 - 55957809 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015120759.2 XP_014976245.1 - 270 165

View Full Product Details