Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHO1
Entrez Gene ID
706145
Gene Name
pleckstrin homology domain containing O1
Gene Aliases
-
Chromosome Location
Chr.1: 101002201 - 101012774 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260966.1 NP_001247895.1 - 207 114
XM_015150590.2 XP_015006076.1 - 713 114
GenBank mRNA JU322623.1 - - 196 114
JU475846.1 - - 446 114
JV044734.1 - - 207 114
JV601706.1 - - 461 114

View Full Product Details