Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CDIPT
Entrez Gene ID
707141
Gene Name
CDP-diacylglycerol--inositol 3-phosphatidyltransferase
Gene Aliases
EGK_12655
Chromosome Location
Chr.20: 27497290 - 27502973 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260643.1 NP_001247572.1 - 573 80
XM_028840493.1 XP_028696326.1 - 1053 80
GenBank mRNA JU321684.1 - - 577 80
JU472849.1 - - 566 80
JV044080.1 - - 573 80
JV567513.1 - - 581 80

View Full Product Details