Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADH6
Entrez Gene ID
707585
Gene Name
alcohol dehydrogenase 6 (class V)
Gene Aliases
-
Chromosome Location
Chr.5: 97497844 - 97514662 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015138866.2 XP_014994352.1 - 382 61

View Full Product Details