Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LASP1
Entrez Gene ID
694950
Gene Name
LIM and SH3 protein 1
Gene Aliases
-
Chromosome Location
Chr.16: 49951426 - 50004567 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265873.1 NP_001252802.1 - 92
XM_028835472.1 XP_028691305.1 - 486 92
XM_028835473.1 XP_028691306.1 - 558 92
GenBank mRNA JU334466.1 - - 711 92
JU470783.1 - - 834 92
JV047038.1 - - 92

View Full Product Details