Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC715706
Entrez Gene ID
715706
Gene Name
schlafen family member 13
Gene Aliases
SLFN13
Chromosome Location
Chr.16: 31792001 - 31802359 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001114211.4 XP_001114211.1 - 1364 75

View Full Product Details