Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN7
Entrez Gene ID
714647
Gene Name
claudin 7
Gene Aliases
-
Chromosome Location
Chr.16: 7027849 - 7030917 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001107265.4 XP_001107265.1 - 821 92
GenBank mRNA JV642621.1 - - 185 92

View Full Product Details