Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTHRC1
Entrez Gene ID
694882
Gene Name
collagen triple helix repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 103648172 - 103660852 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001085750.4 XP_001085750.2 - 315 96
GenBank mRNA JU331146.1 - - 68 96
JV570541.1 - - 161 96
JV605229.1 - - 190 96

View Full Product Details