Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLRC1
Entrez Gene ID
574146
Gene Name
killer cell lectin like receptor C1
Gene Aliases
CD159a, NKG2-A, NKG2A
Chromosome Location
Chr.11: 9270847 - 9280495 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032829.1 NP_001028001.1 - 366 79
XM_028828776.1 XP_028684609.1 - 2538 79
GenBank mRNA AF294891.1 - - 366 79

View Full Product Details