Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTD
Entrez Gene ID
677722
Gene Name
biotinidase
Gene Aliases
-
Chromosome Location
Chr.2: 32021923 - 32067057 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015132222.2 XP_014987708.2 - 603 64
XM_015132223.2 XP_014987709.2 - 523 64
GenBank mRNA AY680553.1 - - 144 64
JU492271.1 - - 564 64
JV659803.1 - - 510 64

View Full Product Details