The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032829.1 | NP_001028001.1 | - | 738 | 91 |
GenBank mRNA | AF190980.1 | - | - | 375 | 91 |
AF190983.1 | - | - | 279 | 91 | |
AF294891.1 | - | - | 738 | 91 | |
AF395618.1 | - | - | 375 | 91 | |
AJ585529.1 | - | - | 354 | 91 |