Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCL25
Entrez Gene ID
574219
Gene Name
C-C motif chemokine ligand 25
Gene Aliases
-
Chromosome Location
Chr.19: 7928413 - 7976670 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032875.1 NP_001028047.1 - 186 71
XM_015122704.2 XP_014978190.1 - 954 71
XM_028838320.1 XP_028694153.1 - 1362 71
XM_028838321.1 XP_028694154.1 - 789 71
XM_028838322.1 XP_028694155.1 - 462 71
XM_028838323.1 XP_028694156.1 - 894 71
XM_028838324.1 XP_028694157.1 - 887 71
XM_028838325.1 XP_028694158.1 - 537 71
GenBank mRNA AF449277.1 - - 186 71
JV658410.1 - - 193 71

View Full Product Details