Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCR3
Entrez Gene ID
715574
Gene Name
natural cytotoxicity triggering receptor 3
Gene Aliases
NKp30
Chromosome Location
Chr.4: 137919086 - 137923382 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY035216.1 - - 224 69

View Full Product Details