Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADGRG1
Entrez Gene ID
613233
Gene Name
adhesion G protein-coupled receptor G1
Gene Aliases
EGK_12829, GPR56
Chromosome Location
Chr.20: 44855566 - 44904737 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033020.1 NP_001028192.1 - 1695 76
XM_015126156.2 XP_014981642.1 - 1861 76
XM_015126157.2 XP_014981643.1 - 2007 76
XM_015126158.2 XP_014981644.1 - 1819 76
XM_015126161.2 XP_014981647.1 - 1819 76
XM_015126162.2 XP_014981648.1 - 1837 76
XM_015126163.2 XP_014981649.1 - 1889 76
XM_015126164.2 XP_014981650.1 - 1858 76
XM_015126165.2 XP_014981651.1 - 1888 76
XM_015126166.2 XP_014981652.1 - 2057 76
XM_015126167.2 XP_014981653.1 - 1804 76
XM_015126168.2 XP_014981654.1 - 1849 76
XM_015126169.2 XP_014981655.1 - 1870 76
XM_015126170.2 XP_014981656.1 - 1821 76
XM_015126171.2 XP_014981657.1 - 2288 76
XM_015126172.2 XP_014981658.1 - 2273 76
XM_028840341.1 XP_028696174.1 - 1752 76
XM_028840342.1 XP_028696175.1 - 1749 76
XM_028840344.1 XP_028696177.1 - 1954 76
XM_028840345.1 XP_028696178.1 - 1736 76
XM_028840346.1 XP_028696179.1 - 1761 76
XM_028840347.1 XP_028696180.1 - 1793 76
GenBank mRNA AY845352.1 - - 1695 76
JU323293.1 - - 1929 76
JU337110.1 - - 1947 76
JU470897.1 - - 1939 76
JU470898.1 - - 1924 76
JU477855.1 - - 1942 76
JV601061.1 - - 1929 76
JV635411.1 - - 1903 76

View Full Product Details