Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADGRG1
Entrez Gene ID
613233
Gene Name
adhesion G protein-coupled receptor G1
Gene Aliases
EGK_12829, GPR56
Chromosome Location
Chr.20: 44855566 - 44904737 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033020.1 NP_001028192.1 - 523 58
XM_015126156.2 XP_014981642.1 - 671 58
XM_015126157.2 XP_014981643.1 - 817 58
XM_015126158.2 XP_014981644.1 - 629 58
XM_015126161.2 XP_014981647.1 - 629 58
XM_015126162.2 XP_014981648.1 - 647 58
XM_015126163.2 XP_014981649.1 - 699 58
XM_015126164.2 XP_014981650.1 - 671 58
XM_015126165.2 XP_014981651.1 - 698 58
XM_015126166.2 XP_014981652.1 - 867 58
XM_015126167.2 XP_014981653.1 - 614 58
XM_015126168.2 XP_014981654.1 - 659 58
XM_015126169.2 XP_014981655.1 - 680 58
XM_015126170.2 XP_014981656.1 - 631 58
XM_015126171.2 XP_014981657.1 - 1116 58
XM_015126172.2 XP_014981658.1 - 1101 58
XM_028840341.1 XP_028696174.1 - 562 58
XM_028840342.1 XP_028696175.1 - 559 58
XM_028840344.1 XP_028696177.1 - 764 58
XM_028840345.1 XP_028696178.1 - 546 58
XM_028840346.1 XP_028696179.1 - 571 58
XM_028840347.1 XP_028696180.1 - 621 58
GenBank mRNA AY845352.1 - - 523 58
JU323293.1 - - 757 58
JU337110.1 - - 757 58
JU470897.1 - - 767 58
JU470898.1 - - 752 58
JU477855.1 - - 752 58
JV601061.1 - - 757 58
JV635411.1 - - 731 58

View Full Product Details