Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LACRT
Entrez Gene ID
654418
Gene Name
lacritin
Gene Aliases
-
Chromosome Location
Chr.11: 54251028 - 54255153 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038659.1 NP_001033748.1 - 117 134
GenBank mRNA DQ191051.1 - - 117 134

View Full Product Details