Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ATRAID
- Entrez Gene ID
-
699201
- Gene Name
- all-trans retinoic acid induced differentiation factor
- Gene Aliases
-
C13H2orf28, C2orf28
- Chromosome Location
-
Chr.13: 81738885 - 81744026 on Build Mmul_10
- Gene Symbol
- SLC5A6
- Entrez Gene ID
-
699074
- Gene Name
- solute carrier family 5 member 6
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 81743815 - 81756819 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV664632.1 |
- |
- |
565 |
79 |
View Full Product Details