Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPE
Entrez Gene ID
708881
Gene Name
carboxypeptidase E
Gene Aliases
-
Chromosome Location
Chr.5: 162794262 - 162914356 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258081.1 NP_001245010.1 - 1373 129
GenBank mRNA JU473276.1 - - 1373 129
JV533763.1 - - 1610 129
JV571640.1 - - 1438 129

View Full Product Details