Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINB1
Entrez Gene ID
707471
Gene Name
serpin family B member 1
Gene Aliases
EGK_14416
Chromosome Location
Chr.4: 167246777 - 167283551 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261583.1 NP_001248512.1 - 472 55
XM_015135409.2 XP_014990895.1 - 682 55
XM_028846756.1 XP_028702589.1 - 691 55
GenBank mRNA JU323205.1 - - 472 55
JU474606.1 - - 473 55
JV044715.1 - - 472 55

View Full Product Details