Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF3E
Entrez Gene ID
697622
Gene Name
eukaryotic translation initiation factor 3 subunit E
Gene Aliases
EIF3S6
Chromosome Location
Chr.8: 108370990 - 108416157 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258088.2 NP_001245017.1 - 867 118
XM_015145867.2 XP_015001353.2 - 976 118
GenBank mRNA JU328028.1 - - 814 118
JU475482.1 - - 871 118
JV044125.1 - - 865 118

View Full Product Details