Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DRG2
Entrez Gene ID
699952
Gene Name
developmentally regulated GTP binding protein 2
Gene Aliases
EGK_08264
Chromosome Location
Chr.16: 17925488 - 17946251 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257836.1 NP_001244765.1 - 684 124
XM_028835501.1 XP_028691334.1 - 762 124
XM_028835502.1 XP_028691335.1 - 762 124
XM_028835503.1 XP_028691336.1 - 672 124
XM_028835504.1 XP_028691337.1 - 672 124
XM_028835505.1 XP_028691338.1 - 672 124
XM_028835506.1 XP_028691339.1 - 775 124
GenBank mRNA JU333254.1 - - 684 124
JU471579.1 - - 690 124
JV046352.1 - - 672 124

View Full Product Details