Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFRSF14
Entrez Gene ID
695042
Gene Name
TNF receptor superfamily member 14
Gene Aliases
-
Chromosome Location
Chr.1: 221832728 - 221839223 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043357.1 NP_001036822.1 - 555 92
GenBank mRNA DQ851094.1 - - 555 92

View Full Product Details