Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAJB6
Entrez Gene ID
693305
Gene Name
DnaJ heat shock protein family (Hsp40) member B6
Gene Aliases
-
Chromosome Location
Chr.3: 183347193 - 183429866 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257911.1 NP_001244840.1 - 503 71
XM_015135376.2 XP_014990862.2 - 548 71
GenBank mRNA JU332386.1 - - 503 71
JU472888.1 - - 415 71
JV044288.1 - - 408 71
JV703311.1 - - 506 71

View Full Product Details