Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLTA
Entrez Gene ID
693659
Gene Name
clathrin light chain A
Gene Aliases
-
Chromosome Location
Chr.15: 47937673 - 47959413 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015117450.1 XP_014972936.1 - 569 66
XM_015117451.1 XP_014972937.1 - 569 66
XM_015117452.1 XP_014972938.1 - 569 66
XM_015117454.1 XP_014972940.1 - 569 66
XM_015117455.1 XP_014972941.1 - 569 66
GenBank mRNA JU320602.1 - - 533 66
JU324557.1 - - 533 66
JU332867.1 - - 533 66
JU332868.1 - - 533 66
JU470478.1 - - 499 66
JU470479.1 - - 499 66
JV046159.1 - - 407 66

View Full Product Details