Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SARAF
Entrez Gene ID
693452
Gene Name
store-operated calcium entry associated regulatory factor
Gene Aliases
EGK_18818, TMEM66
Chromosome Location
Chr.8: 30336055 - 30356717 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015145102.2 XP_015000588.1 - 1614 111
XM_028852526.1 XP_028708359.1 - 1247 111
GenBank mRNA JU322457.1 - - 1134 111
JU475967.1 - - 1136 111
JV045475.1 - - 1027 111
JV638187.1 - - 1132 111

View Full Product Details