Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A11
Entrez Gene ID
710569
Gene Name
solute carrier family 25 member 11
Gene Aliases
-
Chromosome Location
Chr.16: 4784800 - 4787531 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265923.1 NP_001252852.1 - 761 71
XM_015118290.2 XP_014973776.1 - 786 71
GenBank mRNA JU336649.1 - - 778 71
JU477445.1 - - 766 71
JV048410.1 - - 761 71
JV483909.1 - - 386 71

View Full Product Details