Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXCL14
Entrez Gene ID
712076
Gene Name
C-X-C motif chemokine ligand 14
Gene Aliases
-
Chromosome Location
Chr.6: 133032342 - 133040627 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193507.1 NP_001180436.1 - 267 92
GenBank mRNA JU320880.1 - - 217 92
JU473660.1 - - 198 92

View Full Product Details