Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESD
Entrez Gene ID
708871
Gene Name
esterase D
Gene Aliases
-
Chromosome Location
Chr.17: 25219631 - 25247097 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015120908.2 XP_014976394.1 - 206 141
XM_028837139.1 XP_028692972.1 - 585 141
GenBank mRNA JU474655.1 - - 192 141
JV046431.1 - - 206 141
JV459962.1 - - 155 141

View Full Product Details