Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MYL12A
- Entrez Gene ID
-
695077
- Gene Name
- myosin, light chain 12A, regulatory, non-sarcomeric
- Gene Aliases
-
MYL12B
- Chromosome Location
-
Chr.18: 64476464 - 64484961 on Build Mmul_10
- Gene Symbol
- MYL12B
- Entrez Gene ID
-
694957
- Gene Name
- myosin light chain 12B
- Gene Aliases
-
EGK_09550
- Chromosome Location
-
Chr.18: 64490177 - 64506576 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV043393.1 |
- |
- |
826 |
149 |
JV703399.1 |
- |
- |
840 |
149 |
View Full Product Details