Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARGLU1
Entrez Gene ID
702840
Gene Name
arginine and glutamate rich 1
Gene Aliases
-
Chromosome Location
Chr.17: 88095717 - 88120613 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194022.1 NP_001180951.1 - 903 133
GenBank mRNA JU335222.1 - - 762 133
JU335223.1 - - 762 133
JU476185.1 - - 773 133
JU476186.1 - - 773 133
JU476187.1 - - 773 133
JV047495.1 - - 839 133
JV585921.1 - - 745 133
JV636510.1 - - 1083 133
JV688400.1 - - 892 133

View Full Product Details