Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC87
Entrez Gene ID
713677
Gene Name
coiled-coil domain containing 87
Gene Aliases
EGK_05960
Chromosome Location
Chr.14: 7962015 - 7964938 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001109501.4 XP_001109501.2 - 2814 117

View Full Product Details