Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC13
Entrez Gene ID
696521
Gene Name
trafficking protein particle complex 13
Gene Aliases
C6H5orf44
Chromosome Location
Chr.6: 64192859 - 64234750 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266356.1 NP_001253285.1 - 853 151
XM_015140114.2 XP_014995600.1 - 1009 151
XM_015140115.2 XP_014995601.1 - 1297 151
XM_015140116.2 XP_014995602.1 - 991 151
XM_015140117.2 XP_014995603.1 - 736 151
XM_015140118.2 XP_014995604.1 - 777 151
GenBank mRNA JU336462.1 - - 1005 151
JU336463.1 - - 987 151
JU336464.1 - - 987 151
JU336465.1 - - 987 151
JU336466.1 - - 987 151
JU472539.1 - - 966 151
JU472540.1 - - 984 151
JV044090.1 - - 853 151
JV044091.1 - - 835 151

View Full Product Details