Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SYNC
- Entrez Gene ID
-
709783
- Gene Name
- syncoilin, intermediate filament protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 191952954 - 191975907 on Build Mmul_10
- Gene Symbol
- RBBP4
- Entrez Gene ID
-
708710
- Gene Name
- RB binding protein 4, chromatin remodeling factor
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 191975244 - 192004792 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV720369.1 |
- |
- |
3745 |
143 |
View Full Product Details