Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDZD9
Entrez Gene ID
698098
Gene Name
PDZ domain containing 9
Gene Aliases
C20H16orf65, EGK_12583
Chromosome Location
Chr.20: 20241872 - 20259334 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001092453.4 XP_001092453.1 - 107 83

View Full Product Details