Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STX16
Entrez Gene ID
694811
Gene Name
syntaxin 16
Gene Aliases
NPEPL1
Chromosome Location
Chr.10: 93810793 - 93873443 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257651.1 NP_001244580.1 - 932 108
XM_015148665.2 XP_015004151.1 - 284 108
XM_015148666.2 XP_015004152.1 - 362 108
XM_015148667.2 XP_015004153.1 - 248 108
XM_015148668.2 XP_015004154.1 - 289 108
XM_015148670.2 XP_015004156.1 - 284 108
XM_015148671.2 XP_015004157.1 - 311 108
XM_015148672.2 XP_015004158.1 - 253 108
XM_015148673.2 XP_015004159.1 - 284 108
XM_015148674.2 XP_015004160.1 - 227 108
XM_015148675.2 XP_015004161.1 - 233 108
XM_015148676.2 XP_015004162.1 - 129 108
XM_015148681.2 XP_015004167.1 - 284 108
XM_015148682.2 XP_015004168.1 - 362 108
XM_015148683.2 XP_015004169.1 - 392 108
XM_028827168.1 XP_028683001.1 - 289 108
GenBank mRNA JU321093.1 - - 983 108
JU321277.1 - - 920 108
JU321305.1 - - 932 108
JU470546.1 - - 987 108
JU470547.1 - - 936 108
JU470548.1 - - 924 108
JU470549.1 - - 975 108
JV045192.1 - - 936 108
JV045193.1 - - 924 108
JV045194.1 - - 987 108
JV635744.1 - - 975 108
JV638683.1 - - 936 108

View Full Product Details