Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZMAT5
Entrez Gene ID
715058
Gene Name
zinc finger matrin-type 5
Gene Aliases
-
Chromosome Location
Chr.10: 21786212 - 21817748 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194345.1 NP_001181274.1 - 422 98
XM_015150002.2 XP_015005488.1 - 429 98
XM_028827380.1 XP_028683213.1 - 375 98
GenBank mRNA JU339678.1 - - 1090 98
JU473723.1 - - 315 98
JV045029.1 - - 316 98

View Full Product Details