Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIWIL3
Entrez Gene ID
711967
Gene Name
piwi like RNA-mediated gene silencing 3
Gene Aliases
-
Chromosome Location
Chr.10: 26148875 - 26189150 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195743.1 NP_001182672.1 - 363 156
GenBank mRNA HQ025804.1 - - 363 156

View Full Product Details