Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EME2
Entrez Gene ID
722482
Gene Name
essential meiotic structure-specific endonuclease subunit 2
Gene Aliases
-
Chromosome Location
Chr.20: 1796328 - 1799455 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015125310.2 XP_014980796.2 - 975 135
XM_015125311.2 XP_014980797.2 - 1155 135

View Full Product Details