Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB11
Entrez Gene ID
712435
Gene Name
ankyrin repeat and SOCS box containing 11
Gene Aliases
-
Chromosome Location
Chr.X: 14965142 - 14996845 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194155.1 NP_001181084.1 - 256 79
XM_002806154.3 XP_002806200.1 - 289 79

View Full Product Details