Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBLN3
Entrez Gene ID
716357
Gene Name
cerebellin 3 precursor
Gene Aliases
EGK_18073
Chromosome Location
Chr.7: 86515774 - 86521757 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194630.1 NP_001181559.1 - 890 77
GenBank mRNA JV635842.1 - - 831 77

View Full Product Details