Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AADAC
Entrez Gene ID
709031
Gene Name
arylacetamide deacetylase
Gene Aliases
-
Chromosome Location
Chr.2: 54044293 - 54061093 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001106694.4 XP_001106694.1 - 588 181

View Full Product Details