Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR2E1
Entrez Gene ID
701101
Gene Name
nuclear receptor subfamily 2 group E member 1
Gene Aliases
EGK_15469
Chromosome Location
Chr.4: 11861614 - 11883795 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265967.2 NP_001252896.1 - 712 63
GenBank mRNA JU336146.1 - - 714 63
JU336147.1 - - 714 63

View Full Product Details