Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF4
Entrez Gene ID
711169
Gene Name
Kruppel like factor 4
Gene Aliases
-
Chromosome Location
Chr.15: 35397811 - 35402749 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142793.2 NP_001136265.1 - 434 111
XM_028834359.1 XP_028690192.1 - 801 111
GenBank mRNA FJ375752.1 - - 124 111
JV727063.1 - - 434 111

View Full Product Details