Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ENKD1
Entrez Gene ID
699487
Gene Name
enkurin domain containing 1
Gene Aliases
C16orf48, C20H16orf48
Chromosome Location
Chr.20: 54858968 - 54863385 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193959.2 NP_001180888.2 - 1207 150
GenBank mRNA JU325576.1 - - 801 150
JU478347.1 - - 1114 150
JV636959.1 - - 1005 150

View Full Product Details