Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IRX2
Entrez Gene ID
695599
Gene Name
iroquois homeobox 2
Gene Aliases
-
Chromosome Location
Chr.6: 2699478 - 2705363 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015139710.2 XP_014995196.2 - 1732 117
XM_028849326.1 XP_028705159.1 - 1732 117

View Full Product Details