Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC52A3
Entrez Gene ID
715057
Gene Name
solute carrier family 52 member 3
Gene Aliases
C20orf54, EGK_02565
Chromosome Location
Chr.10: 65775491 - 65790948 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194561.2 NP_001181490.1 - 628 105
XM_015149297.2 XP_015004783.1 - 867 105
XM_015149298.2 XP_015004784.1 - 878 105
XM_015149299.2 XP_015004785.1 - 650 105
GenBank mRNA JV046094.1 - - 807 105
JV635392.1 - - 870 105

View Full Product Details