Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM10
Entrez Gene ID
710173
Gene Name
tripartite motif containing 10
Gene Aliases
-
Chromosome Location
Chr.4: 139736419 - 139746177 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127629.1 NP_001121101.1 - 783 117
XM_028846778.1 XP_028702611.1 - 1654 117

View Full Product Details