Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD99L2
Entrez Gene ID
708996
Gene Name
CD99 molecule like 2
Gene Aliases
-
Chromosome Location
Chr.X: 146775340 - 146899380 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261370.1 NP_001248299.1 - 737 82
XM_015128388.2 XP_014983874.1 - 968 82
GenBank mRNA JU336619.1 - - 826 82
JU477432.1 - - 824 82
JV048394.1 - - 737 82
JV630392.1 - - 955 82

View Full Product Details