Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XBP1
Entrez Gene ID
713863
Gene Name
X-box binding protein 1
Gene Aliases
XBP1P1
Chromosome Location
Chr.10: 22747004 - 22754139 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190861.2 NP_001177790.1 - 236 143
NM_001271739.1 NP_001258668.1 - 236 143
GenBank mRNA CO647703.1 - - 240 143
JU336259.1 - - 246 143
JU336260.1 - - 246 143
JV048174.1 - - 243 143
JV627892.1 - - 268 143

View Full Product Details